Gene Symbol: TRNAU1AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017846.4 | NP_060316.1 | 6, 7 | 621 | ||
NR_003109.1 | 7, 8 | 769 | ||||
XM_017001542.1 | XP_016857031.1 | 5, 6 | 795 | |||
GenBank mRNA | AK000510.1 | - | 6, 7 | 553 | ||
BC000680.2 | - | 6, 7 | 545 | 3349210 | ||
BC035855.1 | - | 5, 6 | 1145 | |||
BC039879.1 | - | 7, 8 | 687 | 5769935 |