Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166304.1 | NP_001159776.1 | 3 | 442 | ||
NM_017861.3 | NP_060331.3 | 3 | 442 | |||
GenBank mRNA | AA879458.1 | - | 3 | 212 | ||
AK000529.1 | - | 3 | 405 | |||
AK294340.1 | - | 3 | 625 | |||
BC022542.1 | - | 3 | 402 | 4797783 | ||
BG715890.1 | - | 3 | 448 | |||
CN404010.1 | - | 1 | 118 |