Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271937.1 | NP_001258866.1 | 10 | 893 | ||
NM_017909.3 | NP_060379.2 | 11 | 1411 | |||
XM_005267040.3 | XP_005267097.1 | 10 | 892 | |||
XM_017010988.1 | XP_016866477.1 | 11 | 1410 | |||
GenBank mRNA | AK000634.1 | - | 11 | 1354 | ||
AK026480.1 | - | 10 | 912 | |||
AK292339.1 | - | 11 | 1386 | |||
BC106065.1 | - | 11 | 1373 | 6165385 | ||
BC119683.1 | - | 11 | 1276 | 40116343 | ||
BG754516.1 | - | 1 | 30 |