Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039395.3 | NP_001034484.3 | 6, 7 | 841 | ||
XM_006716795.2 | XP_006716858.1 | 8, 9 | 910 | |||
XM_006716798.3 | XP_006716861.1 | 9, 10 | 915 | |||
XM_006716799.3 | XP_006716862.1 | 7, 8 | 975 | |||
XM_006716800.2 | XP_006716863.1 | 8, 9 | 865 | |||
XM_006716801.2 | XP_006716864.1 | 7, 8 | 966 | |||
XM_011517952.1 | XP_011516254.1 | 8, 9 | 853 | |||
XM_011517954.1 | XP_011516256.1 | 6, 7 | 906 | |||
XM_011517956.1 | XP_011516258.1 | 10, 11 | 1144 | |||
XM_011517957.1 | XP_011516259.1 | 7, 8 | 1036 | |||
XM_011517958.1 | XP_011516260.1 | 8, 9 | 963 | |||
XM_017014881.1 | XP_016870370.1 | 8, 9 | 910 | |||
XM_017014882.1 | XP_016870371.1 | 8, 9 | 910 | |||
XM_017014883.1 | XP_016870372.1 | 8, 9 | 910 | |||
XM_017014884.1 | XP_016870373.1 | 7, 8 | 823 | |||
XM_017014885.1 | XP_016870374.1 | 7, 8 | 670 | |||
XM_017014886.1 | XP_016870375.1 | 6, 7 | 722 | |||
XM_017014887.1 | XP_016870376.1 | 6, 7 | 804 | |||
GenBank mRNA | AK295831.1 | - | 6, 7 | 833 | ||
AK301790.1 | - | 10, 11 | 1244 | |||
BC034293.1 | - | 7, 8 | 942 | 5167647 | ||
BC036349.1 | - | 10, 11 | 1013 |