Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CXorf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184782.1 | NP_001171711.1 | 4, 5 | 1243 | ||
NM_018015.5 | NP_060485.4 | 4, 5 | 1243 | |||
GenBank mRNA | AK001040.1 | - | 4, 5 | 1198 | ||
AK024253.1 | - | 4, 5 | 517 | |||
AK291730.1 | - | 4, 5 | 1192 | |||
BC011483.1 | - | 4, 5 | 1164 | 3453700 | ||
BC033709.1 | - | 4, 5 | 1155 | 3910777 | ||
BC054032.2 | - | 4, 5 | 1131 | |||
BC070110.1 | - | 4, 5 | 1194 | 5264812 |