Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290020.1 | NP_001276949.1 | 13 | 1999 | ||
NM_001290021.1 | NP_001276950.1 | 6 | 825 | |||
NM_001312689.1 | NP_001299618.1 | 8 | 1216 | |||
NM_018076.4 | NP_060546.2 | 13 | 1952 | |||
XM_011519527.1 | XP_011517829.1 | 13 | 1999 | |||
XM_011519528.1 | XP_011517830.1 | 13 | 2649 | |||
XM_011519533.2 | XP_011517835.1 | 8 | 1173 | |||
XM_011519534.2 | XP_011517836.1 | 8 | 1306 | |||
XM_011519535.2 | XP_011517837.1 | 7 | 1058 | |||
XM_011519537.2 | XP_011517839.1 | 4 | 733 | |||
XM_017016368.1 | XP_016871857.1 | 13 | 1999 | |||
XM_017016369.1 | XP_016871858.1 | 13 | 1999 | |||
XM_017016370.1 | XP_016871859.1 | 12 | 1789 | |||
XM_017016371.1 | XP_016871860.1 | 13 | 1999 | |||
GenBank mRNA | AK001679.1 | - | 13 | 1952 | ||
AK292521.1 | - | 13 | 1955 | |||
AK302048.1 | - | 6 | 779 | |||
AK302419.1 | - | 8 | 1548 | |||
AK302627.1 | - | 8 | 1216 | |||
AL136859.1 | - | 2 | 338 | |||
BC140846.1 | - | 12 | 1880 | 9021714 |