Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130447.1 | NP_001123919.1 | 3 | 286 | ||
NM_001323629.1 | NP_001310558.1 | 5 | 468 | |||
NM_001323631.1 | NP_001310560.1 | 5 | 468 | |||
NM_001323632.1 | NP_001310561.1 | 3 | 309 | |||
NM_018097.2 | NP_060567.1 | 4 | 379 | |||
XM_017022396.1 | XP_016877885.1 | 4 | 383 | |||
XM_017022397.1 | XP_016877886.1 | 4 | 350 | |||
GenBank mRNA | AK001322.1 | - | 4 | 337 | ||
AK022939.1 | - | 4 | 350 | |||
BC010903.1 | - | 4 | 358 | 4285311 | ||
DA923763.1 | - | 3 | 243 | |||
DB450370.1 | - | 4 | 379 |