Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175748.3 | NP_786924.2 | 8 | 1134 | ||
NR_038150.1 | 7 | 1000 | ||||
GenBank mRNA | AK001345.1 | - | 5 | 512 | ||
AK097477.1 | - | 8 | 1119 | |||
AK315492.1 | - | 8 | 933 | |||
BC015046.1 | - | 8 | 933 | 3908032 | ||
BC017013.1 | - | 7 | 778 | 3883770 | ||
BC051819.1 | - | 8 | 896 | 6171393 | ||
BX248249.1 | - | 8 | 893 |