Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018122.4 | NP_060592.2 | 10, 11 | 1581 | ||
XM_006711427.3 | XP_006711490.1 | 10, 11 | 1538 | |||
XM_011509711.2 | XP_011508013.1 | 10, 11 | 1538 | |||
GenBank mRNA | AK001376.1 | - | 6, 7 | 585 | ||
AK022754.1 | - | 6, 7 | 586 | |||
AL833633.1 | - | 10, 11 | 1266 | |||
BC045173.2 | - | 10, 11 | 1560 | 5273243 |