Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF10L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011722.2 | NP_001011722.2 | 5 | 422 | ||
NM_001319837.1 | NP_001306766.1 | 5 | 422 | |||
NM_018125.3 | NP_060595.3 | 6 | 538 | |||
NR_137287.1 | 6 | 538 | ||||
NR_137288.1 | 6 | 538 | ||||
XM_005245923.2 | XP_005245980.2 | 6 | 502 | |||
XM_005245925.2 | XP_005245982.2 | 6 | 502 | |||
XM_006710728.1 | XP_006710791.1 | 6 | 684 | |||
XM_006710729.1 | XP_006710792.1 | 6 | 502 | |||
XM_006710731.1 | XP_006710794.1 | 6 | 684 | |||
XM_011541691.2 | XP_011539993.1 | 5 | 508 | |||
XM_011541692.2 | XP_011539994.1 | 5 | 508 | |||
XM_011541693.2 | XP_011539995.1 | 5 | 508 | |||
XM_011541695.2 | XP_011539997.1 | 5 | 508 | |||
XM_011541696.2 | XP_011539998.1 | 5 | 508 | |||
XM_017001617.1 | XP_016857106.1 | 6 | 502 | |||
XM_017001619.1 | XP_016857108.1 | 5 | 508 | |||
XM_017001620.1 | XP_016857109.1 | 5 | 508 | |||
XM_017001621.1 | XP_016857110.1 | 5 | 521 | |||
XM_017001622.1 | XP_016857111.1 | 5 | 521 | |||
GenBank mRNA | AB046846.1 | - | 6 | 538 | ||
AK300174.1 | - | 3 | 186 | |||
AK311076.1 | - | 6 | 465 | |||
BC117171.1 | - | 5 | 412 | 40125722 | ||
BC143347.1 | - | 5 | 412 | 9051852 | ||
DQ139833.1 | - | 5 | 379 |