Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PBRM1 SIRNA Assays ›
Gene Symbol:
PBRM1
Gene Name:
polybromo 1
Gene Aliases:
BAF180, PB1
Chromosome Location:
Chr.3: 52545352 - 52685933 on Build GRCh38
UniGene ID:
Hs.189920
Species:
Human
siRNA ID
HSS124091
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PBRM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_018313.4
NP_060783.3
15, 16
1930
XM_005265279.4
XP_005265336.1
17, 18
2203
XM_005265280.3
XP_005265337.1
16, 17
2020
XM_005265282.3
XP_005265339.1
17, 18
2162
XM_005265283.3
XP_005265340.1
16, 17
2017
XM_011533900.2
XP_011532202.1
19, 20
2369
XM_011533902.2
XP_011532204.1
18, 19
2301
XM_011533903.2
XP_011532205.1
18, 19
2306
XM_017006725.1
XP_016862214.1
18, 19
2276
XM_017006726.1
XP_016862215.1
19, 20
2386
XM_017006727.1
XP_016862216.1
19, 20
2331
XM_017006728.1
XP_016862217.1
18, 19
2327
XM_017006729.1
XP_016862218.1
18, 19
2735
XM_017006730.1
XP_016862219.1
19, 20
2322
XM_017006731.1
XP_016862220.1
18, 19
2305
XM_017006732.1
XP_016862221.1
18, 19
2301
XM_017006733.1
XP_016862222.1
18, 19
2695
XM_017006734.1
XP_016862223.1
18, 19
2268
XM_017006735.1
XP_016862224.1
17, 18
2689
XM_017006736.1
XP_016862225.1
17, 18
2690
XM_017006737.1
XP_016862226.1
17, 18
2609
XM_017006738.1
XP_016862227.1
17, 18
2230
XM_017006739.1
XP_016862228.1
17, 18
2606
XM_017006740.1
XP_016862229.1
17, 18
2147
XM_017006741.1
XP_016862230.1
17, 18
2238
XM_017006742.1
XP_016862231.1
17, 18
2261
XM_017006743.1
XP_016862232.1
18, 19
2735
XM_017006744.1
XP_016862233.1
17, 18
2227
XM_017006745.1
XP_016862234.1
17, 18
2581
XM_017006746.1
XP_016862235.1
17, 18
2578
XM_017006747.1
XP_016862236.1
17, 18
2640
XM_017006748.1
XP_016862237.1
16, 17
2536
XM_017006749.1
XP_016862238.1
16, 17
2102
XM_017006750.1
XP_016862239.1
15, 16
1990
XM_017006751.1
XP_016862240.1
17, 18
2238
XM_017006752.1
XP_016862241.1
18, 19
2735
XM_017006753.1
XP_016862242.1
18, 19
2734
XM_017006754.1
XP_016862243.1
18, 19
2301
XM_017006755.1
XP_016862244.1
17, 18
2238
XM_017006756.1
XP_016862245.1
17, 18
2238
XM_017006757.1
XP_016862246.1
15, 16
1961
XM_017006758.1
XP_016862247.1
15, 16
2092
XM_017006759.1
XP_016862248.1
17, 18
2238
XM_017006760.1
XP_016862249.1
18, 19
2301
XM_017006761.1
XP_016862250.1
18, 19
2310
XM_017006762.1
XP_016862251.1
18, 19
2301
XM_017006763.1
XP_016862252.1
19, 20
2360
XM_017006764.1
XP_016862253.1
17, 18
2238
XM_017006765.1
XP_016862254.1
16, 17
2117
XM_017006766.1
XP_016862255.1
17, 18
2245
GenBank mRNA
AF177387.1
-
14, 15
1819
AF197569.1
-
15, 16
1912
AF225870.1
-
13, 14
1723
AF225871.1
-
14, 15
1819
AF225872.1
-
14, 15
1819
AK056541.1
-
14, 15
1693
AY281068.1
-
14, 15
1818
BC115009.1
-
14, 15
1834
7962003
BC115010.1
-
14, 15
1835
BC115011.1
-
14, 15
1835
7962010
BC129934.1
-
15, 16
1864
40082629
BC129935.1
-
15, 16
1864
40082630
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
negative regulation of cell proliferation
covalent chromatin modification
DNA binding
chromatin binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline