Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NAT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144030.1 | NP_001137502.1 | 7, 8 | 940 | ||
NM_024662.2 | NP_078938.2 | 9, 10 | 1155 | |||
GenBank mRNA | AB051496.1 | - | 9, 10 | 1076 | ||
AF489535.1 | - | 5, 6 | 452 | |||
AK001636.1 | - | 9, 10 | 1010 | |||
AK022241.1 | - | 9, 10 | 1117 | |||
AK225341.1 | - | 9, 10 | 1117 | |||
AK294044.1 | - | 7, 8 | 839 | |||
AL136882.1 | - | 9, 10 | 1012 | |||
BC020205.1 | - | 9, 10 | 977 | |||
BC035558.1 | - | 9, 10 | 1089 | 5166101 |