Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NAT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144030.1 | NP_001137502.1 | 25, 26 | 2907 | ||
NM_024662.2 | NP_078938.2 | 27, 28 | 3122 | |||
GenBank mRNA | AB051496.1 | - | 27, 28 | 3043 | ||
AF489535.1 | - | 23, 24 | 2419 | |||
AK001636.1 | - | 27, 28 | 2977 | |||
AK022241.1 | - | 27, 28 | 3084 | |||
AK225341.1 | - | 27, 28 | 3084 | |||
AK294044.1 | - | 25, 26 | 2806 | |||
AL136882.1 | - | 27, 28 | 2979 | |||
BC035558.1 | - | 27, 28 | 3056 | 5166101 |