Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CASC1 SIRNA Assays ›
Gene Symbol:
CASC1
Gene Name:
cancer susceptibility candidate 1
Gene Aliases:
LAS1, PPP1R54
Chromosome Location:
Chr.12: 25107057 - 25195177 on Build GRCh38
UniGene ID:
Hs.407771
Species:
Human
siRNA ID
HSS124243
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CASC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001082972.2
NP_001076441.1
14
1992
NM_001082973.2
NP_001076442.1
13
1784
NM_001204101.2
NP_001191030.1
11
1607
NM_001204102.2
NP_001191031.1
15
1840
NM_001319977.1
NP_001306906.1
12
1692
NM_001319978.1
NP_001306907.1
13
1958
NM_018272.4
NP_060742.3
14
1802
XM_005253406.1
XP_005253463.1
14
1916
XM_005253408.1
XP_005253465.1
13
1678
XM_005253409.1
XP_005253466.1
12
1692
XM_006719101.1
XP_006719164.1
12
1598
XM_011520723.1
XP_011519025.1
14
1916
XM_017019560.1
XP_016875049.1
13
1881
XM_017019561.1
XP_016875050.1
12
1888
XM_017019562.1
XP_016875051.1
13
1766
XM_017019563.1
XP_016875052.1
12
1480
XM_017019564.1
XP_016875053.1
11
1448
XM_017019565.1
XP_016875054.1
12
1598
XM_017019566.1
XP_016875055.1
12
1809
XM_017019567.1
XP_016875056.1
9
1253
XM_017019568.1
XP_016875057.1
10
1392
GenBank mRNA
AK001783.1
-
8
1206
AK093102.1
-
14
1769
AK297998.1
-
11
1571
AK301763.1
-
12
1353
AK304602.1
-
12
1692
AY423543.1
-
12
1700
BC028415.1
-
14
1933
4823620
BC031047.1
-
14
1783
BC047415.1
-
15
1838
5269578
BC057795.1
-
13
1782
5266518
BC117281.1
-
11
1617
BC117283.1
-
11
1617
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7745c474b-zl6vn:80/100.66.77.21:80.
git-commit: c3c9db9cc82507548abdd0ae6a4ade3f81f66292
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.20.0-2024.11.45-1.0