Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018295.4 | NP_060765.4 | 2 | 332 | ||
GenBank mRNA | AK001862.1 | - | 2 | 274 | ||
AK225055.1 | - | 2 | 262 | |||
AK301268.1 | - | 2 | 262 | |||
BC020942.1 | - | 2 | 259 | 4798940 | ||
DB064371.1 | - | 2 | 333 | |||
DC323016.1 | - | 2 | 332 |
Gene Symbol: C7orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012589.1 | XP_016868078.1 | 4 | 3025 | ||
XM_017012590.1 | XP_016868079.1 | 4 | 2951 | |||
XM_017012591.1 | XP_016868080.1 | 4 | 2805 | |||
XM_017012592.1 | XP_016868081.1 | 3 | 2986 | |||
XM_017012593.1 | XP_016868082.1 | 4 | 2855 | |||
XM_017012594.1 | XP_016868083.1 | 4 | 3062 |