Gene Symbol: STK32B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306082.1 | NP_001293011.1 | 5 | 870 | ||
NM_018401.2 | NP_060871.1 | 4 | 760 | |||
XM_011513497.1 | XP_011511799.1 | 4 | 954 | |||
XM_017008374.1 | XP_016863863.1 | 3 | 361 | |||
XM_017008375.1 | XP_016863864.1 | 3 | 361 | |||
XM_017008376.1 | XP_016863865.1 | 3 | 361 | |||
GenBank mRNA | AJ250839.1 | - | 4 | 477 | ||
AK292696.1 | - | 4 | 727 | |||
AK313846.1 | - | 4 | 762 | |||
AY358353.1 | - | 4 | 504 | |||
BC038238.1 | - | 4 | 454 | 5744969 | ||
BC069203.1 | - | 5 | 791 |