- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: STK32B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306082.1 | NP_001293011.1 | 10, 11 | 1354 | ||
NM_018401.2 | NP_060871.1 | 9, 10 | 1244 | |||
XM_005247982.3 | XP_005248039.1 | 4, 5 | 1751 | |||
XM_005247983.4 | XP_005248040.1 | 4, 5 | 1009 | |||
XM_011513497.1 | XP_011511799.1 | 8, 9 | 1348 | |||
XM_011513501.2 | XP_011511803.1 | 5, 6 | 543 | |||
XM_011513504.1 | XP_011511806.1 | 4, 5 | 397 | |||
XM_017008377.1 | XP_016863866.1 | 4, 5 | 1796 | |||
GenBank mRNA | AJ250839.1 | - | 9, 10 | 961 | ||
AK292696.1 | - | 9, 10 | 1211 | |||
AK313846.1 | - | 9, 10 | 1246 | |||
AL832436.1 | - | 3, 4 | 297 | |||
AY358353.1 | - | 9, 10 | 988 | |||
BC038238.1 | - | 9, 10 | 938 | 5744969 | ||
BC069203.1 | - | 10, 11 | 1275 |