Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LGR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018490.2 | NP_060960.2 | 18 | 3204 | ||
GenBank mRNA | AB209743.1 | - | 18 | 3123 | ||
AF257182.1 | - | 18 | 3163 | |||
BC033039.2 | - | 17 | 3120 | 5260923 |