Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CSGALNACT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319654.1 | NP_001306583.1 | 2 | 1027 | ||
NM_001319656.1 | NP_001306585.1 | 2 | 1027 | |||
NM_018590.4 | NP_061060.3 | 3 | 1150 | |||
NR_135077.1 | 3 | 1150 | ||||
XM_011539938.1 | XP_011538240.1 | 3 | 1150 | |||
XM_017016388.1 | XP_016871877.1 | 3 | 1150 | |||
XM_017016389.1 | XP_016871878.1 | 3 | 1150 | |||
XM_017016390.1 | XP_016871879.1 | 3 | 1150 | |||
XM_017016391.1 | XP_016871880.1 | 3 | 1150 | |||
GenBank mRNA | AB079252.1 | - | 3 | 1081 | ||
AB090811.1 | - | 3 | 1109 | |||
AK125300.1 | - | 3 | 1062 | |||
BC030268.1 | - | 3 | 1109 | 5223190 | ||
BX640967.1 | - | 3 | 1089 | |||
CT003455.1 | - | 2 | 281 | |||
KU178704.1 | - | 2 | 774 | |||
KU178705.1 | - | 2 | 774 | |||
KU178706.1 | - | 2 | 774 |