Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130182.1 | NP_001123654.1 | 2, 3 | 440 | ||
NM_001130183.1 | NP_001123655.1 | 2, 3 | 267 | |||
NM_018602.3 | NP_061072.3 | 3, 4 | 579 | |||
XM_005254535.3 | XP_005254592.1 | 3, 4 | 352 | |||
XM_006720601.1 | XP_006720664.1 | 3, 4 | 354 | |||
GenBank mRNA | AF116663.1 | - | 1, 2 | 671 | ||
AK096616.1 | - | 2, 3 | 440 | |||
AK098079.1 | - | 3, 4 | 570 | |||
BC021720.2 | - | 3, 4 | 579 | 4831235 | ||
BC031044.1 | - | 2, 3 | 302 | 5277834 | ||
BX648710.1 | - | 3, 4 | 461 | |||
DA561365.1 | - | 2, 3 | 268 |