Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPV6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018646.5 | NP_061116.5 | 14, 15 | 2134 | ||
GenBank mRNA | AF304463.1 | - | 14, 15 | 1905 | ||
AF365927.1 | - | 14, 15 | 2121 | |||
AF365928.1 | - | 10, 11 | 1548 | |||
AJ243500.2 | - | 14, 15 | 2105 | |||
AJ243501.2 | - | 14, 15 | 2105 | |||
AJ487964.2 | - | 15, 16 | 2074 | |||
AK075103.1 | - | 8, 9 | 1635 | |||
AK075206.1 | - | 4, 5 | 504 | |||
AK291707.1 | - | 14, 15 | 2135 | |||
AY203947.1 | - | 11, 12 | 2315 | |||
KF534785.1 | - | 14, 15 | 2134 |