Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130037.1 | NP_001123509.1 | 7 | 885 | ||
NM_001308018.1 | NP_001294947.1 | 8 | 1021 | |||
NM_018712.3 | NP_061182.3 | 7 | 885 | |||
XM_017017994.1 | XP_016873483.1 | 8 | 1016 | |||
XM_017017995.1 | XP_016873484.1 | 7 | 766 | |||
GenBank mRNA | AK054794.1 | - | 7 | 705 | ||
AK297351.1 | - | 8 | 1021 | |||
AK303689.1 | - | 7 | 727 | |||
AL359601.1 | - | 1 | 72 | |||
BC028725.2 | - | 7 | 628 | 4815682 |