Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017625.2 | NP_060095.2 | 7 | 825 | ||
GenBank mRNA | AB036706.1 | - | 7 | 737 | ||
AF271386.1 | - | 7 | 748 | |||
AK000029.1 | - | 7 | 774 | |||
AK222533.1 | - | 7 | 812 | |||
AY065972.1 | - | 7 | 815 | |||
AY157361.1 | - | 7 | 827 | |||
AY157362.1 | - | 7 | 819 | |||
AY358359.1 | - | 7 | 811 | |||
AY549722.1 | - | 7 | 914 | |||
AY619691.1 | - | 7 | 735 | |||
AY619692.1 | - | 7 | 792 | |||
BC020664.1 | - | 7 | 825 | 4693329 | ||
CR457224.1 | - | 6 | 709 |