Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THUMPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304550.1 | NP_001291479.1 | 3 | 654 | ||
NM_017736.4 | NP_060206.2 | 3 | 654 | |||
XM_011545891.1 | XP_011544193.1 | 4 | 438 | |||
XM_017023433.1 | XP_016878922.1 | 3 | 596 | |||
GenBank mRNA | AK000281.1 | - | 3 | 508 | ||
AK294328.1 | - | 1 | 87 | |||
BC000448.2 | - | 3 | 486 | 2821354 | ||
BX640898.1 | - | 3 | 774 |