Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017797.3 | NP_060267.2 | 4 | 727 | ||
XM_005259593.1 | XP_005259650.1 | 4 | 492 | |||
XM_011528127.1 | XP_011526429.1 | 2 | 93 | |||
GenBank mRNA | AF355797.1 | - | 4 | 727 | ||
AK000393.1 | - | 1 | 521 | |||
AK027481.1 | - | 2 | 89 | |||
AK225831.1 | - | 4 | 726 | |||
AY007116.1 | - | 4 | 449 | |||
BC000564.2 | - | 1 | 17 | |||
BC008035.2 | - | 11 | ||||
BC069199.1 | - | 2 | 153 | |||
BC094820.1 | - | 4 | 461 |