Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC37L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017913.3 | NP_060383.2 | 2 | 420 | ||
GenBank mRNA | AK000497.1 | - | 2 | 415 | ||
AK000646.1 | - | 2 | 304 | |||
AK225510.1 | - | 2 | 304 | |||
BC014133.2 | - | 2 | 240 | 4622495 | ||
DA180237.1 | - | 2 | 420 |