Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KANSL3 SIRNA Assays ›
Gene Symbol:
KANSL3
Gene Name:
KAT8 regulatory NSL complex subunit 3
Gene Aliases:
KIAA1310, NSL3, Rcd1
Chromosome Location:
Chr.2: 96593155 - 96638379 on Build GRCh38
UniGene ID:
Hs.516341
Species:
Human
siRNA ID
HSS124798
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KANSL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001115016.2
NP_001108488.1
8
1116
NR_047653.1
8
1177
NR_047654.1
8
1177
NR_047655.1
7
1025
NR_047656.1
7
1025
NR_047657.1
8
1116
NR_047658.1
8
1093
XM_005263982.2
XP_005264039.1
8
1115
XM_005263984.1
XP_005264041.1
8
1115
XM_005263987.1
XP_005264044.1
8
1115
XM_011511454.2
XP_011509756.1
8
1115
XM_011511455.2
XP_011509757.1
8
1084
XM_011511456.2
XP_011509758.1
8
1118
XM_011511457.2
XP_011509759.1
8
1037
XM_011511458.2
XP_011509760.1
8
1115
XM_011511459.2
XP_011509761.1
8
1115
XM_011511460.2
XP_011509762.1
8
1115
XM_011511461.2
XP_011509763.1
8
1115
XM_011511462.2
XP_011509764.1
7
944
XM_011511463.2
XP_011509765.1
8
1128
XM_011511464.2
XP_011509766.1
8
1187
XM_011511466.2
XP_011509768.1
8
1128
XM_011511467.2
XP_011509769.1
7
970
XM_017004470.1
XP_016859959.1
8
1115
XM_017004471.1
XP_016859960.1
7
1022
XM_017004472.1
XP_016859961.1
8
1128
XM_017004473.1
XP_016859962.1
8
1128
XM_017004474.1
XP_016859963.1
8
1023
GenBank mRNA
AB037731.1
-
8
1068
AF311326.1
-
10
929
AK000943.1
-
6
1017
AK023813.1
-
8
1144
AK302646.1
-
7
966
AK304011.1
-
7
1025
AK311617.1
-
7
958
AK314367.1
-
8
1053
AY050169.1
-
8
1144
BC015469.2
-
9
1110
BC024276.1
-
8
1053
BC032746.1
-
8
1187
BC051763.1
-
6
747
BC063792.1
-
8
1046
6504387
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
histone H4-K5 acetylation
histone H4-K8 acetylation
histone H4-K16 acetylation
histone acetyltransferase activity (H4-K5 specific)
histone acetyltransferase activity (H4-K8 specific)
histone acetyltransferase activity (H4-K16 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-n6vvh:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0