Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KANSL3 SIRNA Assays ›
Gene Symbol:
KANSL3
Gene Name:
KAT8 regulatory NSL complex subunit 3
Gene Aliases:
KIAA1310, NSL3, Rcd1
Chromosome Location:
Chr.2: 96593155 - 96638379 on Build GRCh38
UniGene ID:
Hs.516341
Species:
Human
siRNA ID
HSS124800
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KANSL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001115016.2
NP_001108488.1
19
2608
NR_047653.1
19
2669
NR_047654.1
19
2663
NR_047655.1
19
2595
NR_047656.1
18
2517
XM_005263982.2
XP_005264039.1
20
2685
XM_005263984.1
XP_005264041.1
20
2685
XM_005263987.1
XP_005264044.1
19
2601
XM_011511454.2
XP_011509756.1
20
2685
XM_011511455.2
XP_011509757.1
20
2654
XM_011511456.2
XP_011509758.1
20
2688
XM_011511457.2
XP_011509759.1
20
2607
XM_011511458.2
XP_011509760.1
20
2679
XM_011511459.2
XP_011509761.1
19
2610
XM_011511460.2
XP_011509762.1
19
2607
XM_011511461.2
XP_011509763.1
19
2601
XM_011511462.2
XP_011509764.1
19
2514
XM_011511463.2
XP_011509765.1
20
2698
XM_011511464.2
XP_011509766.1
20
2757
XM_011511466.2
XP_011509768.1
19
2620
XM_011511467.2
XP_011509769.1
19
2540
XM_017004470.1
XP_016859959.1
20
2679
XM_017004471.1
XP_016859960.1
19
2592
XM_017004472.1
XP_016859961.1
20
2692
XM_017004473.1
XP_016859962.1
20
2698
XM_017004474.1
XP_016859963.1
20
2593
GenBank mRNA
AF311326.1
-
21
2415
AK023813.1
-
19
2630
AK302646.1
-
19
2536
AK304011.1
-
18
2517
AK314367.1
-
19
2545
AL136849.1
-
5
609
AY050169.1
-
19
2636
BC051763.1
-
18
2317
BC063792.1
-
19
2538
6504387
BQ063147.1
-
5
553
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
histone H4-K5 acetylation
histone H4-K8 acetylation
histone H4-K16 acetylation
histone acetyltransferase activity (H4-K5 specific)
histone acetyltransferase activity (H4-K8 specific)
histone acetyltransferase activity (H4-K16 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0