Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RADIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018059.4 | NP_060529.4 | 8 | 2086 | ||
GenBank mRNA | AB058752.1 | - | 8 | 2030 | ||
AK000997.1 | - | 4 | 524 | |||
AK001186.1 | - | 5 | 1069 | |||
AK294745.1 | - | 7 | 1885 | |||
AK296325.1 | - | 7 | 1607 | |||
AK302549.1 | - | 2078 | ||||
AL136731.1 | - | 3 | 317 | |||
BC117317.1 | - | 7 | 1972 | 40125868 | ||
BC126311.1 | - | 7 | 1972 | 8992027 | ||
BC143526.1 | - | 7 | 1972 | 9052035 | ||
EF560723.1 | - | 7 | 1995 |