Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018154.2 | NP_060624.1 | 2 | 317 | ||
GenBank mRNA | AB104486.1 | - | 2 | 145 | ||
AF279307.1 | - | 2 | 238 | |||
AK001288.1 | - | 2 | 276 | |||
AK001466.1 | - | 2 | 308 | |||
AK223080.1 | - | 2 | 271 | |||
AK302134.1 | - | 2 | 278 | |||
BC007726.2 | - | 2 | 241 | 4303318 | ||
BC010014.2 | - | 2 | 251 | 4304756 | ||
BC036521.1 | - | 2 | 281 | 5270216 | ||
CN426445.1 | - | 2 | 652 | |||
CR457235.1 | - | 2 | 145 |