Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ST6GALNAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289107.1 | NP_001276036.1 | 9, 10 | 1940 | ||
NM_018414.4 | NP_060884.1 | 8, 9 | 1803 | |||
NR_110309.1 | 7, 8 | 1699 | ||||
GenBank mRNA | AK000113.1 | - | 7, 8 | 957 | ||
AY096001.1 | - | 8, 9 | 1603 | |||
AY358918.1 | - | 8, 9 | 1748 | |||
BC022462.1 | - | 7, 8 | 1554 | 4816713 | ||
Y11339.2 | - | 8, 9 | 1663 |