Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRERF1 SIRNA Assays ›
Gene Symbol:
TRERF1
Gene Name:
transcriptional regulating factor 1
Gene Aliases:
BCAR2, HSA277276, RAPA, TREP132, TReP-132, dJ139D8.5
Chromosome Location:
Chr.6: 42224933 - 42452216 on Build GRCh38
UniGene ID:
Hs.485392
Species:
Human
siRNA ID
HSS125085
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRERF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001297573.1
NP_001284502.1
14
3298
NM_033502.3
NP_277037.1
14
3238
XM_011514741.2
XP_011513043.1
15
3935
XM_011514742.2
XP_011513044.1
17
4111
XM_011514743.2
XP_011513045.1
16
4034
XM_011514744.1
XP_011513046.1
15
3543
XM_011514745.1
XP_011513047.1
15
3437
XM_017011034.1
XP_016866523.1
15
3985
XM_017011035.1
XP_016866524.1
15
3653
XM_017011036.1
XP_016866525.1
15
3472
XM_017011037.1
XP_016866526.1
15
3503
XM_017011038.1
XP_016866527.1
17
4125
XM_017011039.1
XP_016866528.1
15
3730
XM_017011040.1
XP_016866529.1
15
3422
XM_017011041.1
XP_016866530.1
15
3544
XM_017011042.1
XP_016866531.1
14
4647
XM_017011043.1
XP_016866532.1
14
5657
XM_017011044.1
XP_016866533.1
15
3934
XM_017011045.1
XP_016866534.1
15
3874
XM_017011046.1
XP_016866535.1
14
3716
XM_017011047.1
XP_016866536.1
15
3796
XM_017011048.1
XP_016866537.1
14
3716
XM_017011049.1
XP_016866538.1
14
3656
XM_017011050.1
XP_016866539.1
15
3935
XM_017011051.1
XP_016866540.1
14
3533
XM_017011052.1
XP_016866541.1
13
3407
XM_017011053.1
XP_016866542.1
15
3583
XM_017011054.1
XP_016866543.1
13
3407
XM_017011055.1
XP_016866544.1
14
2241
GenBank mRNA
AF297872.1
-
14
3232
AJ277275.1
-
13
2773
AJ277276.1
-
13
2773
AK127493.1
-
10
2712
AM404182.1
-
14
3298
AM404183.1
-
14
3238
AM404259.1
-
14
3238
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
steroid biosynthetic process
cholesterol catabolic process
multicellular organism development
homeostatic process
positive regulation of transcription, DNA-templated
regulation of hormone biosynthetic process
RNA polymerase II transcription cofactor activity
transcription factor activity, sequence-specific DNA binding
transcription factor binding
DNA binding, bending
ligand-dependent nuclear receptor transcription coactivator activity
transcription regulatory region DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-f4tdd:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0