Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TSNAXIP1 SIRNA Assays ›
Gene Symbol:
TSNAXIP1
Gene Name:
translin associated factor X interacting protein 1
Gene Aliases:
TXI1
Chromosome Location:
Chr.16: 67806573 - 67828068 on Build GRCh38
UniGene ID:
Hs.632212
Species:
Human
siRNA ID
HSS125101
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TSNAXIP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001288990.1
NP_001275919.1
10
1532
NM_001288991.1
NP_001275920.1
8
1325
NM_001288992.1
NP_001275921.1
6
1278
NM_001288993.1
NP_001275922.1
7
1405
NM_001288994.1
NP_001275923.1
5
1081
NM_018430.3
NP_060900.2
10
1721
XM_005256049.4
XP_005256106.1
10
1726
XM_011523230.2
XP_011521532.1
9
1933
XM_011523231.2
XP_011521533.1
9
1912
XM_011523232.2
XP_011521534.1
10
1885
XM_011523233.2
XP_011521535.1
10
1858
XM_011523234.2
XP_011521536.1
9
1801
XM_011523235.1
XP_011521537.1
9
1568
XM_011523236.2
XP_011521538.1
7
1191
XM_011523237.2
XP_011521539.1
7
1693
XM_011523238.1
XP_011521540.1
6
1106
XM_011523239.1
XP_011521541.1
7
1107
XM_011523240.1
XP_011521542.1
8
1293
XM_011523241.1
XP_011521543.1
7
1381
XM_011523242.1
XP_011521544.1
6
1234
XM_011523243.1
XP_011521545.1
6
1188
XM_017023445.1
XP_016878934.1
7
1170
XM_017023446.1
XP_016878935.1
7
1037
XM_017023447.1
XP_016878936.1
7
1059
XM_017023448.1
XP_016878937.1
7
1010
XM_017023449.1
XP_016878938.1
8
984
XM_017023450.1
XP_016878939.1
9
1197
XM_017023451.1
XP_016878940.1
9
1383
XM_017023452.1
XP_016878941.1
6
1213
XM_017023453.1
XP_016878942.1
8
1285
XM_017023454.1
XP_016878943.1
7
1138
XM_017023455.1
XP_016878944.1
7
1027
GenBank mRNA
AF132730.1
-
7
1209
AK027245.1
-
10
1439
AK301920.1
-
10
1305
AK302300.1
-
714
AK303841.1
-
8
1115
BC111018.1
-
10
1475
5168384
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
spermatogenesis
biological_process
cell differentiation
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0