Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TSNAXIP1 SIRNA Assays ›
Gene Symbol:
TSNAXIP1
Gene Name:
translin associated factor X interacting protein 1
Gene Aliases:
TXI1
Chromosome Location:
Chr.16: 67806573 - 67828068 on Build GRCh38
UniGene ID:
Hs.632212
Species:
Human
siRNA ID
HSS125102
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TSNAXIP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001288990.1
NP_001275919.1
12
1744
NM_001288991.1
NP_001275920.1
10
1537
NM_001288992.1
NP_001275921.1
8
1490
NM_001288993.1
NP_001275922.1
9
1617
NM_001288994.1
NP_001275923.1
7
1293
NM_018430.3
NP_060900.2
12
1933
XM_005256049.4
XP_005256106.1
12
1938
XM_011523230.2
XP_011521532.1
11
2145
XM_011523231.2
XP_011521533.1
11
2124
XM_011523232.2
XP_011521534.1
12
2097
XM_011523233.2
XP_011521535.1
12
2070
XM_011523234.2
XP_011521536.1
11
2013
XM_011523235.1
XP_011521537.1
11
1780
XM_011523236.2
XP_011521538.1
9
1403
XM_011523237.2
XP_011521539.1
9
1905
XM_011523238.1
XP_011521540.1
8
1318
XM_011523239.1
XP_011521541.1
9
1319
XM_011523240.1
XP_011521542.1
10
1505
XM_011523241.1
XP_011521543.1
9
1593
XM_011523242.1
XP_011521544.1
8
1446
XM_011523243.1
XP_011521545.1
8
1400
XM_017023445.1
XP_016878934.1
9
1382
XM_017023446.1
XP_016878935.1
9
1249
XM_017023447.1
XP_016878936.1
9
1271
XM_017023448.1
XP_016878937.1
9
1222
XM_017023449.1
XP_016878938.1
10
1196
XM_017023450.1
XP_016878939.1
11
1409
XM_017023451.1
XP_016878940.1
11
1595
XM_017023452.1
XP_016878941.1
8
1425
XM_017023453.1
XP_016878942.1
10
1497
XM_017023454.1
XP_016878943.1
9
1350
XM_017023455.1
XP_016878944.1
9
1239
GenBank mRNA
AF132730.1
-
9
1421
AK027245.1
-
12
1651
AK301920.1
-
12
1517
AK302300.1
-
926
AK303841.1
-
10
1327
BC111018.1
-
12
1687
5168384
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
spermatogenesis
biological_process
cell differentiation
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-dvnl5:80/10.239.162.83:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0