Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GLT8D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010983.2 | NP_001010983.1 | 7 | 1363 | ||
NM_001278280.1 | NP_001265209.1 | 7 | 1359 | |||
NM_001278281.1 | NP_001265210.1 | 7 | 1107 | |||
NM_018446.3 | NP_060916.1 | 6 | 1134 | |||
NM_152932.2 | NP_690909.1 | 6 | 692 | |||
XM_017006857.1 | XP_016862346.1 | 8 | 1522 | |||
XM_017006858.1 | XP_016862347.1 | 7 | 1293 | |||
XM_017006859.1 | XP_016862348.1 | 6 | 842 | |||
GenBank mRNA | AF157318.1 | - | 6 | 579 | ||
AF175227.1 | - | 7 | 846 | |||
AJ271710.1 | - | 5 | 461 | |||
AK027517.1 | - | 6 | 600 | |||
AK075031.1 | - | 5 | 656 | |||
AY358579.1 | - | 6 | 613 | |||
BC119670.1 | - | 6 | 578 | 40116196 | ||
BC119671.1 | - | 6 | 578 | 40116197 | ||
CR749821.1 | - | 7 | 816 |