Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282529.1 | NP_001269458.1 | 8 | 767 | ||
NM_018449.3 | NP_060919.3 | 12 | 1081 | |||
GenBank mRNA | AK026088.1 | - | 4 | 249 | ||
AK294952.1 | - | 12 | 999 | |||
AK299183.1 | - | 6 | 489 | |||
AK302335.1 | - | 10 | 902 | |||
AK302754.1 | - | 11 | 949 | |||
AK303493.1 | - | 8 | 705 | |||
BC111762.1 | - | 6 | 451 | |||
BF332025.1 | - | 1 | 94 |