Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282529.1 | NP_001269458.1 | 12 | 1636 | ||
NM_018449.3 | NP_060919.3 | 16 | 1950 | |||
GenBank mRNA | AB040924.1 | - | 4 | 752 | ||
AK025801.1 | - | 5 | 840 | |||
AK026088.1 | - | 8 | 1118 | |||
AK074884.1 | - | 3 | 309 | |||
AK294952.1 | - | 16 | 1868 | |||
AK299183.1 | - | 10 | 1358 | |||
AK302335.1 | - | 14 | 1771 | |||
AK302754.1 | - | 15 | 1818 | |||
AK303493.1 | - | 12 | 1574 | |||
AY358682.1 | - | 4 | 1160 | |||
BC111762.1 | - | 10 | 1320 |