Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018660.2 | NP_061130.1 | 2 | 300 | ||
GenBank mRNA | AB044750.1 | - | 1 | 123 | ||
AB057659.1 | - | 2 | 306 | |||
AB073627.1 | - | 2 | 306 | |||
AF263928.1 | - | 3 | 510 | |||
AK002050.1 | - | 2 | 225 | |||
AK023418.1 | - | 2 | 300 | |||
AK098243.1 | - | 8 | 1183 | |||
BC001237.1 | - | 2 | 262 | 3452822 |