Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019116.2 | NP_061989.2 | 2 | 539 | ||
XM_011545894.2 | XP_011544196.1 | 2 | 545 | |||
XM_017023459.1 | XP_016878948.1 | 2 | 545 | |||
XM_017023460.1 | XP_016878949.1 | 2 | 545 | |||
GenBank mRNA | AK092696.1 | - | 2 | 353 | ||
AK124139.1 | - | 3 | 315 |