Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291277.1 | NP_001278206.1 | 3 | 1186 | ||
NM_031274.4 | NP_112564.1 | 3 | 1212 | |||
GenBank mRNA | AF285597.1 | - | 3 | 1193 | ||
BC042547.1 | - | 3 | 1167 | 5164310 | ||
BC108734.1 | - | 3 | 1185 | 5744723 |