Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019592.6 | NP_062538.5 | 3 | 364 | ||
XM_011518862.1 | XP_011517164.1 | 3 | 641 | |||
GenBank mRNA | AF265230.1 | - | 3 | 362 | ||
AK000389.1 | - | 2 | 255 | |||
AK000697.1 | - | 2 | 255 | |||
AK022532.1 | - | 3 | 360 | |||
AK314401.1 | - | 3 | 319 | |||
BC016691.1 | - | 4 | 319 | |||
BC063115.1 | - | 3 | 345 | |||
BC110584.1 | - | 3 | 357 | 40009044 | ||
BC110585.1 | - | 3 | 357 | 40009047 | ||
BC152309.1 | - | 3 | 357 | 40009034 | ||
DB273543.1 | - | 3 | 364 |