Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020643.2 | NP_065694.2 | 2 | 151 | ||
XM_017018013.1 | XP_016873502.1 | 1 | 612 | |||
GenBank mRNA | AK074455.1 | - | 2 | 246 | ||
AK223377.1 | - | 2 | 246 | |||
BC027865.1 | - | 2 | 133 | 5203029 | ||
DA399415.1 | - | 2 | 151 |