Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317830.1 | NP_001304759.1 | 4 | 1850 | ||
NM_020647.3 | NP_065698.1 | 4 | 1850 | |||
XM_005251274.3 | XP_005251331.1 | 4 | 2034 | |||
XM_005251275.4 | XP_005251332.1 | 4 | 2034 | |||
GenBank mRNA | BC049372.1 | - | 4 | 1461 | ||
BC098299.1 | - | 4 | 1814 | |||
BC098314.1 | - | 4 | 1451 | |||
BC099736.1 | - | 4 | 1451 | |||
BC113856.1 | - | 4 | 1816 | |||
BC114464.1 | - | 4 | 1816 | |||
BC139832.1 | - | 4 | 1812 | |||
BC140875.1 | - | 4 | 1850 | 9021743 | ||
BC140876.1 | - | 4 | 1850 | 9021744 | ||
BX537756.1 | - | 4 | 1716 | |||
CR627477.1 | - | 1 | 152 |