Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKS1B SIRNA Assays ›
Gene Symbol:
ANKS1B
Gene Name:
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases:
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
Chromosome Location:
Chr.12: 98729904 - 99984727 on Build GRCh38
UniGene ID:
Hs.506458
Species:
Human
siRNA ID
HSS125693
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKS1B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204065.1
NP_001190994.1
6
800
NM_001204066.1
NP_001190995.1
7
667
NM_001204067.1
NP_001190996.1
7
761
NM_001204068.1
NP_001190997.1
7
1224
NM_001204069.1
NP_001190998.1
7
1116
NM_001204070.1
NP_001190999.1
6
1044
NM_001204079.1
NP_001191008.1
6
803
NM_001204080.1
NP_001191009.1
5
728
NM_001204081.1
NP_001191010.1
5
728
NM_020140.3
NP_064525.1
6
1044
NM_152788.4
NP_690001.3
21
3656
NM_181670.3
NP_858056.2
8
1296
XM_005269028.4
XP_005269085.1
21
3267
XM_005269029.4
XP_005269086.1
21
3227
XM_005269032.3
XP_005269089.1
5
592
XM_006719504.3
XP_006719567.1
22
3801
XM_006719505.3
XP_006719568.1
22
3804
XM_006719506.3
XP_006719569.1
22
3801
XM_006719507.3
XP_006719570.1
22
3804
XM_006719508.3
XP_006719571.1
22
3804
XM_006719509.3
XP_006719572.1
21
3729
XM_006719510.3
XP_006719573.1
21
3729
XM_006719512.3
XP_006719575.1
20
3549
XM_006719513.3
XP_006719576.1
20
3549
XM_006719514.3
XP_006719577.1
20
3537
XM_011538571.2
XP_011536873.1
22
3702
XM_017019651.1
XP_016875140.1
22
3804
XM_017019652.1
XP_016875141.1
22
3792
XM_017019653.1
XP_016875142.1
22
3801
XM_017019654.1
XP_016875143.1
22
3789
XM_017019655.1
XP_016875144.1
22
3801
XM_017019656.1
XP_016875145.1
22
3699
XM_017019657.1
XP_016875146.1
21
3627
XM_017019658.1
XP_016875147.1
20
3549
XM_017019659.1
XP_016875148.1
8
1295
XM_017019660.1
XP_016875149.1
7
1220
XM_017019661.1
XP_016875150.1
9
1006
XM_017019662.1
XP_016875151.1
9
1139
XM_017019663.1
XP_016875152.1
8
1190
XM_017019664.1
XP_016875153.1
8
876
XM_017019665.1
XP_016875154.1
6
1040
GenBank mRNA
AF145204.1
-
15
2360
AF164792.1
-
6
977
AK289768.1
-
8
1261
AK294191.1
-
7
761
AK294994.1
-
5
600
AK295988.1
-
6
773
AK315954.1
-
6
655
AM980945.1
-
6
659
AY281131.1
-
21
3242
AY281132.1
-
7
917
AY283057.2
-
13
2093
AY356353.1
-
5
557
AY620824.1
-
12
1967
AY753193.1
-
6
737
BC026313.1
-
8
1191
BC068451.1
-
7
667
BC142669.1
-
6
991
BC150204.1
-
7
1063
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of synaptic plasticity by receptor localization to synapse
ephrin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-sshrn:80/10.239.165.44:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0