Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020158.3 | NP_064543.3 | 2 | 245 | ||
GenBank mRNA | AF281134.1 | - | 2 | 245 | ||
AF285785.1 | - | 2 | 376 | |||
AK314941.1 | - | 2 | 230 | |||
BC007742.2 | - | 2 | 228 | 4308795 | ||
BC107696.1 | - | 2 | 250 | 6453881 |