Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PARP6 SIRNA Assays ›
Gene Symbol:
PARP6
Gene Name:
poly(ADP-ribose) polymerase family member 6
Gene Aliases:
ARTD17, PARP-6-B1, PARP-6-C, pART17
Chromosome Location:
Chr.15: 72241181 - 72272575 on Build GRCh38
UniGene ID:
Hs.270244
Species:
Human
siRNA ID
HSS125831
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PARP6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001323515.1
NP_001310444.1
9
948
NM_001323516.1
NP_001310445.1
9
1021
NM_001323519.1
NP_001310448.1
9
1070
NM_001323521.1
NP_001310450.1
9
1015
NM_001323522.1
NP_001310451.1
9
960
NM_001323523.1
NP_001310452.1
9
1243
NM_001323524.1
NP_001310453.1
9
1021
NM_001323525.1
NP_001310454.1
9
1131
NM_001323526.1
NP_001310455.1
9
1131
NM_001323528.1
NP_001310457.1
9
1021
NM_001323530.1
NP_001310459.1
9
1131
NM_001323531.1
NP_001310460.1
9
960
NM_001323532.1
NP_001310461.1
9
1131
NM_020214.3
NP_064599.2
9
1021
NR_136594.1
9
1131
NR_136596.1
9
1274
NR_136599.1
9
1021
NR_136603.1
9
1015
NR_136604.1
9
1131
NR_136605.1
9
1131
NR_136606.1
9
1243
NR_136607.1
9
1131
NR_136608.1
9
1021
NR_136609.1
9
1131
NR_136610.1
9
1131
NR_136611.1
9
1131
XM_011521805.1
XP_011520107.1
4
393
XM_017022418.1
XP_016877907.1
8
1671
GenBank mRNA
AB499727.1
-
6
489
AB499728.1
-
6
489
AK021790.1
-
7
583
AK022547.1
-
8
712
AK126416.1
-
6
1314
AL122091.1
-
4
303
AL390087.1
-
6
692
BC015314.1
-
5
347
BC026955.1
-
5
346
BC110902.1
-
7
537
6194444
BC119688.2
-
7
548
BC119689.1
-
7
548
Back To Top
More Information
Gene Ontology Categories:
Process(es)
NAD+ ADP-ribosyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline