Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020198.2 | NP_064583.2 | 10 | 1390 | ||
XM_005257527.2 | XP_005257584.1 | 10 | 1258 | |||
GenBank mRNA | AF113221.1 | - | 8 | 737 | ||
AF226054.1 | - | 10 | 1238 | |||
AK027786.1 | - | 10 | 1232 | |||
AK027844.1 | - | 10 | 1219 | |||
AK075387.1 | - | 11 | 1319 | |||
AK225813.1 | - | 10 | 1258 | |||
AK314328.1 | - | 10 | 1245 | |||
BC008905.2 | - | 10 | 1212 | 3688068 | ||
BC013600.2 | - | 7 | 681 |