Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020312.3 | NP_064708.1 | 7, 8 | 951 | ||
GenBank mRNA | AF161444.1 | - | 7, 8 | 885 | ||
AK290627.1 | - | 7, 8 | 881 | |||
AL136884.1 | - | 2, 3 | 385 | |||
BC001478.2 | - | 5, 6 | 526 | |||
BC029341.1 | - | 7, 8 | 885 | |||
BC054340.1 | - | 7, 8 | 885 | 6514554 | ||
BC064946.1 | - | 7, 8 | 886 | 6023461 |