Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300743.1 | NP_001287672.1 | 8, 9 | 1029 | ||
NM_020314.5 | NP_064710.4 | 8, 9 | 1029 | |||
XM_005255435.3 | XP_005255492.1 | 8, 9 | 981 | |||
XM_006721065.3 | XP_006721128.1 | 8, 9 | 981 | |||
XM_011545896.2 | XP_011544198.1 | 7, 8 | 659 | |||
GenBank mRNA | AK024693.1 | - | 4, 5 | 299 | ||
AK095078.1 | - | 8, 9 | 726 | |||
AK290286.1 | - | 8, 9 | 726 | |||
AK294885.1 | - | 9, 10 | 1084 | |||
AL832159.1 | - | 8, 9 | 725 | |||
AL833428.1 | - | 8, 9 | 718 | |||
BC014900.2 | - | 7, 8 | 697 | 3909068 | ||
BC050464.1 | - | 8, 9 | 769 | 6061139 | ||
BC058845.1 | - | 8, 9 | 728 | 6452778 | ||
BX537867.1 | - | 8, 9 | 741 |