Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLSCR2 SIRNA Assays ›
Gene Symbol:
PLSCR2
Gene Name:
phospholipid scramblase 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 146391363 - 146497163 on Build GRCh38
UniGene ID:
Hs.744414
Species:
Human
siRNA ID
HSS125924
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLSCR2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199978.1
NP_001186907.1
8
1187
NM_001199979.1
NP_001186908.1
6
804
NM_020359.2
NP_065092.1
7
917
XM_011513013.2
XP_011511315.2
6
1967
XM_011513019.2
XP_011511321.2
6
1967
XM_011513020.2
XP_011511322.2
6
1967
XM_011513021.2
XP_011511323.2
6
1967
XM_011513022.2
XP_011511324.2
6
1967
XM_011513023.2
XP_011511325.2
6
1967
XM_017006898.1
XP_016862387.1
6
1967
XM_017006899.1
XP_016862388.1
6
1967
XM_017006900.1
XP_016862389.1
6
1967
XM_017006901.1
XP_016862390.1
6
1967
XM_017006902.1
XP_016862391.1
6
1967
XM_017006903.1
XP_016862392.1
6
1967
XM_017006904.1
XP_016862393.1
6
1967
XM_017006905.1
XP_016862394.1
8
1423
XM_017006906.1
XP_016862395.1
8
1204
XM_017006907.1
XP_016862396.1
8
969
XM_017006908.1
XP_016862397.1
8
1094
XM_017006909.1
XP_016862398.1
7
881
XM_017006910.1
XP_016862399.1
6
994
XM_017006911.1
XP_016862400.1
7
1271
XM_017006912.1
XP_016862401.1
6
622
XM_017006913.1
XP_016862402.1
4
1593
XM_017006914.1
XP_016862403.1
6
829
XM_017006915.1
XP_016862404.1
6
811
GenBank mRNA
AA813518.1
-
3
411
AF159441.1
-
7
861
AK301909.1
-
8
1187
BC055415.1
-
6
808
BC069785.1
-
6
754
7262547
BC120969.2
-
6
826
40115378
BC141969.1
-
6
826
40115468
Back To Top
More Information
Panther Classification:
Molecular Function -
transporter
Gene Ontology Categories:
Function(s)
Process(es)
phospholipid scrambling
calcium ion binding
phospholipid scramblase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-b9vj9:80/10.239.164.80:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0